ENA375227355 Enamine

1-(3-methoxyphenyl)-2-methylpiperazine dihydrochloride

  • MDL number MFCD22565898
  • Cas Number : 1423025-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236...238
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114565

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114565
Availability details for
Quantity
  • Availability
  • Add to Cart