ENA375227358 Enamine

3-amino-2-methoxypropan-1-ol hydrochloride

  • MDL number MFCD22565901
  • Cas Number : 1423028-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114607
Availability details for
Quantity
  • Availability
  • Add to Cart