ENA375227365 Enamine

2-cyclopropyl-2-(dimethylamino)acetic acid hydrochloride

  • MDL number MFCD22565908
  • Cas Number : 1423024-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114643
Availability details for
Quantity
  • Availability
  • Add to Cart