ENA375227371 Enamine

5H,6H,7H,8H-imidazo[1,5-a]pyridine-7-carboxylic acid hydrochloride

  • MDL number MFCD22565914
  • Cas Number : 180718-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114688
Availability details for
Quantity
  • Availability
  • Add to Cart