ENA375227373 Enamine

4-amino-2,2-dimethyl-3-phenylbutan-1-ol

  • MDL number MFCD22565916
  • Cas Number : 1423025-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114695
Availability details for
Quantity
  • Availability
  • Add to Cart