ENA375227378 Enamine

(1R)-7-methoxy-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number MFCD22565921
  • Cas Number : 1423040-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114736
Availability details for
Quantity
  • Availability
  • Add to Cart