ENA375227379 Enamine

methyl (3S)-3-amino-3-(3,5-dichlorophenyl)propanoate hydrochloride

  • MDL number MFCD22565922
  • Cas Number : 1423040-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114738
Availability details for
Quantity
  • Availability
  • Add to Cart