ENA375227381 Enamine

N-[(3-bromoadamantan-1-yl)methyl]-2-chloroacetamide

  • Cas Number : 1423026-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114742
Availability details for
Quantity
  • Availability
  • Add to Cart