ENA375227382 Enamine

(3-bromoadamantan-1-yl)methanamine hydrochloride

  • Cas Number : 895167-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 299...301
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114747
Availability details for
Quantity
  • Availability
  • Add to Cart