ENA375227383 Enamine

(1-benzyl-5-methylpyrrolidin-3-yl)methanamine

  • MDL number MFCD22565926
  • Cas Number : 1423031-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114750
Availability details for
Quantity
  • Availability
  • Add to Cart