ENA375227393 Enamine

methyl 6-(piperazin-1-yl)pyridazine-3-carboxylate dihydrochloride

  • MDL number MFCD22565937
  • Cas Number : 1423025-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114857

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114857
Availability details for
Quantity
  • Availability
  • Add to Cart