ENA375227400 Enamine

3-[(2-fluorophenyl)methyl]pyrrolidin-3-ol hydrochloride

  • MDL number MFCD22565944
  • Cas Number : 1246635-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-114914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114914
Availability details for
Quantity
  • Availability
  • Add to Cart