ENA375227402 Enamine

2-amino-3-(oxan-2-yl)propanoic acid hydrochloride

  • MDL number MFCD22565946
  • Cas Number : 1423026-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114945
Availability details for
Quantity
  • Availability
  • Add to Cart