ENA375227404 Enamine

2-(1-chloroethyl)-5-(trifluoromethyl)-1,3,4-oxadiazole

  • MDL number MFCD22565949
  • Cas Number : 762241-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114963
Availability details for
Quantity
  • Availability
  • Add to Cart