ENA375227409 Enamine

1-aminocyclobutane-1-carboxamide hydrochloride

  • MDL number MFCD22565954
  • Cas Number : 190004-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115058

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115058
Availability details for
Quantity
  • Availability
  • Add to Cart