ENA375227419 Enamine

methyl 2,6-dichloro-3,5-dinitrobenzoate

  • MDL number MFCD22565968
  • Cas Number : 1146-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115214

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115214
Availability details for
Quantity
  • Availability
  • Add to Cart