ENA375227422 Enamine

2-[ethyl(methyl)amino]-2-methylpropanoic acid hydrochloride

  • MDL number MFCD22565971
  • Cas Number : 1423031-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115239

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115239
Availability details for
Quantity
  • Availability
  • Add to Cart