ENA375227423 Enamine

5-chloro-3-(methylamino)-2,3-dihydro-1$l^{6}-benzothiophene-1,1-dione hydrochloride

  • MDL number MFCD22565972
  • Cas Number : 1423034-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115244

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115244
Availability details for
Quantity
  • Availability
  • Add to Cart