ENA375227424 Enamine

4-ethenyl-1-(2-methoxyethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD22565973
  • Cas Number : 1423033-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115250

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115250
Availability details for
Quantity
  • Availability
  • Add to Cart