ENA375227425 Enamine

7-amino-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one hydrochloride

  • MDL number MFCD22565975
  • Cas Number : 1423026-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115301
Availability details for
Quantity
  • Availability
  • Add to Cart