ENA375227430 Enamine

6-fluoro-2-(piperidin-3-yl)-1,3-benzoxazole hydrochloride

  • MDL number MFCD22565980
  • Cas Number : 1423024-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115414

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115414
Availability details for
Quantity
  • Availability
  • Add to Cart