ENA375227444 Enamine

2-amino-N-[(2-fluorophenyl)methyl]-N-methylpentanamide hydrochloride

  • MDL number MFCD22566000
  • Cas Number : 1423024-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115605

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115605
Availability details for
Quantity
  • Availability
  • Add to Cart