ENA375227446 Enamine

methyl 2-(aminomethyl)-2-ethylbutanoate hydrochloride

  • MDL number MFCD22566002
  • Cas Number : 177269-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115622

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115622
Availability details for
Quantity
  • Availability
  • Add to Cart