ENA375227447 Enamine

N-(oxan-4-yl)-1,2,3,4-tetrahydroisoquinoline-3-carboxamide hydrochloride

  • MDL number MFCD22566004
  • Cas Number : 1423024-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115640
Availability details for
Quantity
  • Availability
  • Add to Cart