ENA375227462 Enamine

2-amino-N-methyl-2-phenyl-N-[(thiophen-3-yl)methyl]acetamide hydrochloride

  • MDL number MFCD22566020
  • Cas Number : 1423024-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115767
Availability details for
Quantity
  • Availability
  • Add to Cart