ENA375227477 Enamine

2-({[(tert-butoxy)carbonyl]amino}methyl)-2-ethylbutanoic acid

  • MDL number MFCD22566037
  • Cas Number : 204514-21-6
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-115932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115932
Availability details for
Quantity
  • Availability
  • Add to Cart