ENA375227478 Enamine

5-chloro-2-(chloromethyl)-4-methoxypyridine hydrochloride

  • MDL number MFCD22566038
  • Cas Number : 122307-30-6
  • Molecular Weight : 228.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-(chloromethyl)-4-methoxypyridine hydrochloride
Inchi Code 1S/C7H7Cl2NO.ClH/c1-11-7-2-5(3-8)10-4-6(7)9;/h2,4H,3H2,1H3;1H
Inchi Key NNDAYCLMPFWHFS-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-115948

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-115948
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart