ENA375227480 Enamine

2-amino-N-(3,4-dihydro-2H-1-benzothiopyran-4-yl)pentanamide hydrochloride

  • MDL number MFCD22566040
  • Cas Number : 1423024-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115950

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115950
Availability details for
Quantity
  • Availability
  • Add to Cart