ENA375227482 Enamine

2-amino-N-(6-chloro-3,4-dihydro-2H-1-benzothiopyran-4-yl)pentanamide hydrochloride

  • MDL number MFCD22566042
  • Cas Number : 1423024-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-115952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115952
Availability details for
Quantity
  • Availability
  • Add to Cart