ENA375227484 Enamine

3-(aminomethyl)-2,5-dihydro-1,2,4-oxadiazol-5-one hydrochloride

  • MDL number MFCD22566044
  • Cas Number : 1423025-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116002
Availability details for
Quantity
  • Availability
  • Add to Cart