ENA375227487 Enamine

cyclohexyl (2S)-2-aminobutanoate hydrochloride

  • MDL number MFCD22566047
  • Cas Number : 37666-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116043
Availability details for
Quantity
  • Availability
  • Add to Cart