ENA375227488 Enamine

N''-(piperazine-1-carboximidoyl)guanidine trihydrochloride

  • MDL number MFCD22566049
  • Cas Number : 1423024-26-3
  • Molecular Weight : 279.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N''-[imino(1-piperazinyl)methyl]guanidine trihydrochloride
Inchi Code 1S/C6H14N6.3ClH/c7-5(8)11-6(9)12-3-1-10-2-4-12;;;/h10H,1-4H2,(H5,7,8,9,11);3*1H
Inchi Key GKWFWTUOSWZEPE-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-116100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116100
Availability details for
Quantity
  • Availability
  • Add to Cart