ENA375227490 Enamine

1,1-dimethyl-3-[2-(methylamino)ethyl]urea hydrochloride

  • MDL number MFCD22566052
  • Cas Number : 1423033-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116131
Availability details for
Quantity
  • Availability
  • Add to Cart