ENA375227501 Enamine

N-cyclopropyl-2-(thiomorpholin-3-yl)-N-[(thiophen-3-yl)methyl]acetamide hydrochloride

  • MDL number MFCD22566064
  • Cas Number : 1423034-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116224
Availability details for
Quantity
  • Availability
  • Add to Cart