ENA375227518 Enamine

1-[2-(2-chlorophenoxy)ethyl]-6-oxo-1,6-dihydropyridine-2-carboxylic acid

  • MDL number MFCD22566085
  • Cas Number : 1423031-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116533

Safety Information

msds https://www.enaminestore.com/catalog/EN300-116533
Availability details for
Quantity
  • Availability
  • Add to Cart