ENA375227523 Enamine

(4-methoxyphenyl)(pyrrolidin-2-yl)methanol

  • MDL number MFCD07390357
  • Cas Number : 1139238-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114629
Availability details for
Quantity
  • Availability
  • Add to Cart