ENA375227526 Enamine

2-(cyclopentylamino)-N-(2,6-dichlorophenyl)acetamide

  • MDL number MFCD12563291
  • Cas Number : 1156724-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114633
Availability details for
Quantity
  • Availability
  • Add to Cart