ENA375227529 Enamine

3-amino-1-phenylbutan-2-one hydrochloride

  • MDL number MFCD20134012
  • Cas Number : 191104-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114637

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114637
Availability details for
Quantity
  • Availability
  • Add to Cart