ENA375227538 Enamine

4-chloro-6-methoxy-2-methylquinazoline

  • MDL number MFCD12024901
  • Cas Number : 60395-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114654
Availability details for
Quantity
  • Availability
  • Add to Cart