ENA375227541 Enamine

2-bromoquinoline-3-carbonitrile

  • MDL number MFCD20296193
  • Cas Number : 1000007-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114657
Availability details for
Quantity
  • Availability
  • Add to Cart