ENA375227544 Enamine

ethyl 3-(4-bromophenyl)-3-hydroxypropanoate

  • MDL number MFCD16749036
  • Cas Number : 70200-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-112174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112174
Availability details for
Quantity
  • Availability
  • Add to Cart