ENA375227547 Enamine

1-(bromomethyl)-3-(methoxymethyl)benzene

  • MDL number MFCD15523515
  • Cas Number : 125604-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114682

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114682
Availability details for
Quantity
  • Availability
  • Add to Cart