ENA375227553 Enamine

2-(methylsulfanyl)butanoic acid

  • MDL number MFCD12823560
  • Cas Number : 874525-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114694
Availability details for
Quantity
  • Availability
  • Add to Cart