ENA375227560 Enamine

4-[(1-methyl-1H-1,2,4-triazol-5-yl)methoxy]benzene-1-sulfonyl chloride

  • MDL number MFCD21267959
  • Cas Number : 1423031-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114710

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114710
Availability details for
Quantity
  • Availability
  • Add to Cart