ENA375227567 Enamine

4,7-difluoro-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD11219516
  • Cas Number : 130408-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114723
Availability details for
Quantity
  • Availability
  • Add to Cart