ENA375227572 Enamine

1-(3-methoxy-4-sulfanylphenyl)ethan-1-one

  • MDL number MFCD22565846
  • Cas Number : 69114-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104744
Availability details for
Quantity
  • Availability
  • Add to Cart