ENA375227575 Enamine

1-{[1,2,3,4]tetrazolo[1,5-a]pyrazin-5-yl}-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD16531313
  • Cas Number : 1282434-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113009
Availability details for
Quantity
  • Availability
  • Add to Cart