ENA375227589 Enamine

3-amino-2-chloropyridine-4-carboxylic acid

  • MDL number MFCD15143308
  • Cas Number : 58483-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114788

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-114788
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart