ENA375227596 Enamine

2-(3-chlorophenoxy)-4-methylbenzoic acid

  • MDL number MFCD17378694
  • Cas Number : 1273836-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113330
Availability details for
Quantity
  • Availability
  • Add to Cart