ENA375227600 Enamine

1-(4-ethoxyphenyl)piperazine dihydrochloride

  • MDL number MFCD12913780
  • Cas Number : 53689-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221...223
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114802

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-114802
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart